Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAGGGCCGTCAGACTGTCGA[A/G]CCTAAGAATAAAAAGACCCTGTTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614288 MIM: 610552 | |||||||||||||||||||||||
Literature Links: |
ACAD11 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACAD11 - acyl-CoA dehydrogenase family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032169.4 | 657 | Intron | NP_115545.3 |
NPHP3-ACAD11 - NPHP3-ACAD11 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBA5 - ubiquitin like modifier activating enzyme 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320210.1 | 657 | Intron | NP_001307139.1 | |||
NM_001321238.1 | 657 | Intron | NP_001308167.1 | |||
NM_001321239.1 | 657 | UTR 5 | NP_001308168.1 | |||
NM_024818.4 | 657 | UTR 5 | NP_079094.1 | |||
NM_198329.3 | 657 | Intron | NP_938143.1 |