Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCAGTCCAGGCCGGATCCCAG[C/T]AGCCGTTCCCTTTCCTTTCTTCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF485 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF485 - zinc finger protein 485 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318140.1 | Intron | NP_001305069.1 | ||||
NM_001318141.1 | Intron | NP_001305070.1 | ||||
NM_001318142.1 | Intron | NP_001305071.1 | ||||
NM_001318143.1 | Intron | NP_001305072.1 | ||||
NM_145312.3 | Intron | NP_660355.2 | ||||
XM_011539498.2 | Intron | XP_011537800.1 |
Set Membership: |
HapMap |