Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCGGCGGCTGCAGCGGCAGCAG[A/C]TGCTGCCCTGCACCCGGCGCTCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613483 MIM: 603711 | ||||||||||||||||||||
Literature Links: |
BHLHE22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BHLHE22 - basic helix-loop-helix family member e22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152414.4 | 1545 | Missense Mutation | GAT,GCT | D,A 331 | NP_689627.1 |
CYP7B1 - cytochrome P450 family 7 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC401463 - uncharacterized LOC401463 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |