Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCAAAATGTAAATGATTCAGCT[A/C]ACTGAGTTCATGTCTAATTAGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613272 MIM: 608400 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCTD3 - potassium channel tetramerization domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USH2A - usherin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007123.5 | 18098 | Intron | NP_009054.5 | |||
NM_206933.2 | 18098 | UTR 3 | NP_996816.2 |