Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCCTGACCAGCCAGGCCCAGCG[C/G]TTCCCCGCCTACTGCATTTGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606247 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAMBP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STAMBP - STAM binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006463.4 | 189 | Intron | NP_006454.1 | |||
NM_201647.2 | 189 | Intron | NP_964010.1 | |||
NM_213622.2 | 189 | UTR 5 | NP_998787.1 | |||
XM_005264088.3 | 189 | Intron | XP_005264145.1 | |||
XM_011532483.2 | 189 | Intron | XP_011530785.1 | |||
XM_011532484.2 | 189 | Intron | XP_011530786.1 | |||
XM_011532485.2 | 189 | Intron | XP_011530787.1 | |||
XM_017003175.1 | 189 | Intron | XP_016858664.1 | |||
XM_017003176.1 | 189 | Intron | XP_016858665.1 | |||
XM_017003177.1 | 189 | Intron | XP_016858666.1 | |||
XM_017003178.1 | 189 | UTR 5 | XP_016858667.1 | |||
XM_017003179.1 | 189 | Intron | XP_016858668.1 | |||
XM_017003180.1 | 189 | Intron | XP_016858669.1 | |||
XM_017003181.1 | 189 | UTR 5 | XP_016858670.1 |
Set Membership: |
HapMap |