Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATTATCAGGAAGTATATTTCAA[C/T]GATTAAATTTTCACCTTCCAAGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603201 MIM: 608058 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCB11 - ATP binding cassette subfamily B member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003742.2 | Intron | NP_003733.2 | ||||
XM_006712817.3 | Intron | XP_006712880.1 | ||||
XM_011512077.2 | Intron | XP_011510379.1 | ||||
XM_011512078.2 | Intron | XP_011510380.1 | ||||
XM_011512080.2 | Intron | XP_011510382.1 | ||||
XM_011512081.2 | Intron | XP_011510383.1 | ||||
XM_017005165.1 | Intron | XP_016860654.1 | ||||
XM_017005166.1 | Intron | XP_016860655.1 | ||||
XM_017005167.1 | Intron | XP_016860656.1 |
G6PC2 - glucose-6-phosphatase catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |