Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGAGGATGAAGCAAAGTTCTAC[C/T]GGATTATAAATGGTCTGATCTATTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602705 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYN3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SYN3 - synapsin III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135774.1 | 4852 | Intron | NP_001129246.1 | |||
NM_003490.3 | 4852 | Intron | NP_003481.3 | |||
NM_133633.2 | 4852 | Intron | NP_598344.2 | |||
XM_011530405.2 | 4852 | UTR 3 | XP_011528707.1 | |||
XM_011530406.2 | 4852 | UTR 3 | XP_011528708.1 | |||
XM_011530407.2 | 4852 | UTR 3 | XP_011528709.1 | |||
XM_011530408.1 | 4852 | UTR 3 | XP_011528710.1 | |||
XM_011530410.2 | 4852 | UTR 3 | XP_011528712.1 | |||
XM_017028960.1 | 4852 | UTR 3 | XP_016884449.1 | |||
XM_017028961.1 | 4852 | UTR 3 | XP_016884450.1 | |||
XM_017028962.1 | 4852 | UTR 3 | XP_016884451.1 | |||
XM_017028963.1 | 4852 | UTR 3 | XP_016884452.1 | |||
XM_017028964.1 | 4852 | UTR 3 | XP_016884453.1 | |||
XM_017028965.1 | 4852 | UTR 3 | XP_016884454.1 |
Set Membership: |
HapMap |