Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCTATTCTTTTCCTCGTTTTGC[A/T]CTGGCATGCCTTCATTTTCAACACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612754 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRX3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLRX3 - glutaredoxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199868.1 | Intron | NP_001186797.1 | ||||
NM_001321980.1 | Intron | NP_001308909.1 | ||||
NM_006541.4 | Intron | NP_006532.2 | ||||
XM_017015474.1 | Intron | XP_016870963.1 |
Set Membership: |
HapMap |