Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTTGAAACAGGGCTTACTAAA[A/G]TCTCCTTGGATAAGACTCAACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610769 MIM: 608414 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOC3L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NOC3L - NOC3 like DNA replication regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022451.10 | Intron | NP_071896.8 | ||||
XM_017016544.1 | Intron | XP_016872033.1 |
PLCE1 - phospholipase C epsilon 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |