Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATGCTGTTCTTGGTCTTCCAC[A/G]AGAAAGTGAAGCTGTCAGCTTAATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611505 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPP - centromere protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012267.2 | 1761 | UTR 3 | NP_001012267.1 | |||
NM_001286969.1 | 1761 | UTR 3 | NP_001273898.1 | |||
NM_001286971.1 | 1761 | UTR 3 | NP_001273900.1 | |||
XM_011518685.2 | 1761 | Intron | XP_011516987.1 | |||
XM_011518689.1 | 1761 | Intron | XP_011516991.1 | |||
XM_017014715.1 | 1761 | Intron | XP_016870204.1 |
IPPK - inositol-pentakisphosphate 2-kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022755.5 | 1761 | UTR 3 | NP_073592.1 | |||
XM_017015041.1 | 1761 | Intron | XP_016870530.1 |
LOC100128361 - uncharacterized LOC100128361 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |