Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCAAGAGTTTGTAACTTGGTAC[A/C]ATGTGCCTTTATTATTTTCTGCATC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602648 | |||||||||||||||||||||||
Literature Links: |
ACKR2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACKR2 - atypical chemokine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001296.4 | Intron | NP_001287.2 |
HIGD1A - HIG1 hypoxia inducible domain family member 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |