Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCTGCATTTGAAGTTTGCAGAG[G/T]ATCCACTTTTCCTCTTTTTGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605439 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EHF - ETS homologous factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206615.1 | Intron | NP_001193544.1 | ||||
NM_001206616.1 | Intron | NP_001193545.1 | ||||
NM_012153.5 | Intron | NP_036285.2 | ||||
XM_005252857.1 | Intron | XP_005252914.1 | ||||
XM_005252860.1 | Intron | XP_005252917.1 | ||||
XM_005252861.1 | Intron | XP_005252918.1 | ||||
XM_005252862.1 | Intron | XP_005252919.1 | ||||
XM_011519983.1 | Intron | XP_011518285.1 | ||||
XM_011519984.1 | Intron | XP_011518286.1 | ||||
XM_011519985.1 | Intron | XP_011518287.1 | ||||
XM_017017534.1 | Intron | XP_016873023.1 |
Set Membership: |
HapMap |