Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACTCAGCTCGTCGACCTGCACG[C/T]CCGATACGCTGAGTGTGGTTTGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138322 MIM: 615729 | ||||||||||||||||||||||||||||||||
Literature Links: |
GPX4 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GPX4 - glutathione peroxidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039847.2 | 459 | Missense Mutation | GCC,GTC | A,V 88 | NP_001034936.1 | |
NM_001039848.2 | 459 | Missense Mutation | GCC,GTC | A,V 125 | NP_001034937.1 | |
NM_002085.4 | 459 | Missense Mutation | GCC,GTC | A,V 88 | NP_002076.2 |
SBNO2 - strawberry notch homolog 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
DME Validated Inventoried |