Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGACAGAATCAAGCTATACTGG[A/G]GGAAAACGTTGCTTCTCTAGAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612088 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC12A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC12A - C-type lectin domain family 12 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207010.1 | Intron | NP_001193939.1 | ||||
NM_001300730.1 | Intron | NP_001287659.1 | ||||
NM_138337.5 | Intron | NP_612210.4 | ||||
NM_201623.3 | Intron | NP_963917.2 | ||||
XM_005253324.2 | Intron | XP_005253381.1 | ||||
XM_006719033.3 | Intron | XP_006719096.1 | ||||
XM_006719035.3 | Intron | XP_006719098.1 | ||||
XM_006719036.3 | Intron | XP_006719099.1 | ||||
XM_011520570.1 | Intron | XP_011518872.1 | ||||
XM_011520571.2 | Intron | XP_011518873.2 | ||||
XM_011520573.2 | Intron | XP_011518875.1 |
LOC400002 - uncharacterized LOC400002 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |