Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAATTCAAAAGTGGTCTCAAATT[A/G]TGAACAGGCAGATAACACCTAGCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602769 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNMT3A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNMT3A - DNA methyltransferase 3 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320892.1 | Intron | NP_001307821.1 | ||||
NM_001320893.1 | Intron | NP_001307822.1 | ||||
NM_022552.4 | Intron | NP_072046.2 | ||||
NM_153759.3 | Intron | NP_715640.2 | ||||
NM_175629.2 | Intron | NP_783328.1 | ||||
NM_175630.1 | Intron | NP_783329.1 | ||||
XM_005264175.4 | Intron | XP_005264232.1 | ||||
XM_005264177.4 | Intron | XP_005264234.1 | ||||
XM_011532662.2 | Intron | XP_011530964.1 | ||||
XM_011532663.2 | Intron | XP_011530965.1 | ||||
XM_011532664.2 | Intron | XP_011530966.1 | ||||
XM_011532666.2 | Intron | XP_011530968.1 | ||||
XM_011532667.2 | Intron | XP_011530969.1 | ||||
XM_017003526.1 | Intron | XP_016859015.1 | ||||
XM_017003527.1 | Intron | XP_016859016.1 |
Set Membership: |
HapMap |