Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAAATTAAAGCAGAAAACTGTC[C/T]AGGAAAATCAGACTTCAAAGTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107470 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNGR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFNGR1 - interferon gamma receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000416.2 | 1685 | UTR 3 | NP_000407.1 | |||
XM_006715470.2 | 1685 | UTR 3 | XP_006715533.1 | |||
XM_006715471.2 | 1685 | UTR 3 | XP_006715534.1 | |||
XM_011535793.2 | 1685 | UTR 3 | XP_011534095.1 | |||
XM_011535794.1 | 1685 | UTR 3 | XP_011534096.1 | |||
XM_017010827.1 | 1685 | UTR 3 | XP_016866316.1 |