Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610480 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEMD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LEMD1 - LEM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001552.4 | Intron | NP_001001552.3 | ||||
NM_001199050.1 | Intron | NP_001185979.1 | ||||
NM_001199051.1 | Intron | NP_001185980.1 | ||||
NM_001199052.1 | Intron | NP_001185981.1 | ||||
XM_011510160.2 | Intron | XP_011508462.1 | ||||
XM_011510161.2 | Intron | XP_011508463.1 | ||||
XM_011510162.2 | Intron | XP_011508464.1 | ||||
XM_011510163.2 | Intron | XP_011508465.1 | ||||
XM_011510164.2 | Intron | XP_011508466.1 | ||||
XM_011510165.2 | Intron | XP_011508467.1 |
LEMD1-AS1 - LEMD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |