Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAACCACAAAAGCAAATGTGGTC[A/T]TGTTTCTGATTTTAGTTAAGTCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GK5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GK5 - glycerol kinase 5 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039547.2 | 4282 | UTR 3 | NP_001034636.1 | |||
XM_011512624.2 | 4282 | UTR 3 | XP_011510926.1 | |||
XM_017006088.1 | 4282 | Intron | XP_016861577.1 | |||
XM_017006089.1 | 4282 | UTR 3 | XP_016861578.1 | |||
XM_017006090.1 | 4282 | UTR 3 | XP_016861579.1 |