Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAAGTTGGGGTTTTCCATTGCAG[A/C]TGCCACTCGCTCTTTGTCATTTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM96A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM96A - family with sequence similarity 96 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014812.2 | 661 | UTR 3 | NP_001014812.1 | |||
NM_001289108.1 | 661 | UTR 3 | NP_001276037.1 | |||
NM_032231.5 | 661 | Missense Mutation | GCT,TCT | A,S 141 | NP_115607.1 |