Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155760 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACAN - aggrecan | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135.3 | Intron | NP_001126.3 | ||||
NM_013227.3 | Intron | NP_037359.3 | ||||
XM_006720419.1 | Intron | XP_006720482.1 | ||||
XM_011521313.1 | Intron | XP_011519615.1 | ||||
XM_011521314.1 | Intron | XP_011519616.1 | ||||
XM_017021985.1 | Intron | XP_016877474.1 | ||||
XM_017021986.1 | Intron | XP_016877475.1 | ||||
XM_017021987.1 | Intron | XP_016877476.1 |
Set Membership: |
HapMap |