Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTCAAGATTTTCTCTTTATCTT[G/T]GGTTTTCAATAGTTTGTGTCTAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
YIPF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YIPF6 - Yip1 domain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195214.1 | Intron | NP_001182143.1 | ||||
NM_173834.3 | Intron | NP_776195.2 | ||||
XM_011530923.2 | Intron | XP_011529225.1 | ||||
XM_011530924.1 | Intron | XP_011529226.1 | ||||
XM_017029450.1 | Intron | XP_016884939.1 | ||||
XM_017029451.1 | Intron | XP_016884940.1 |