Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120620 | ||||||||||||||||||||
Literature Links: |
CR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CR1 - complement component 3b/4b receptor 1 (Knops blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000573.3 | 460 | Missense Mutation | GCG,GTG | A,V 115 | NP_000564.2 | |
NM_000651.4 | 460 | Missense Mutation | GCG,GTG | A,V 115 | NP_000642.3 | |
XM_006711166.2 | 460 | Missense Mutation | GCG,GTG | A,V 120 | XP_006711229.1 | |
XM_011509205.1 | 460 | Missense Mutation | GCG,GTG | A,V 120 | XP_011507507.1 |