Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607709 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TJP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TJP2 - tight junction protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170414.2 | Intron | NP_001163885.1 | ||||
NM_001170415.1 | Intron | NP_001163886.1 | ||||
NM_001170416.1 | Intron | NP_001163887.1 | ||||
NM_004817.3 | Intron | NP_004808.2 | ||||
NM_201629.3 | Intron | NP_963923.1 | ||||
XM_005252314.2 | Intron | XP_005252371.1 | ||||
XM_011519204.1 | Intron | XP_011517506.1 | ||||
XM_011519206.2 | Intron | XP_011517508.1 | ||||
XM_011519207.2 | Intron | XP_011517509.1 | ||||
XM_011519208.2 | Intron | XP_011517510.1 | ||||
XM_011519209.2 | Intron | XP_011517511.1 | ||||
XM_017015326.1 | Intron | XP_016870815.1 | ||||
XM_017015327.1 | Intron | XP_016870816.1 | ||||
XM_017015328.1 | Intron | XP_016870817.1 |