Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGGAATCCGAAGTTGGAACCCC[C/G]GGGGGCCGAGGGCTGCCGGTCCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ID1 - inhibitor of DNA binding 1, HLH protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002165.3 | 459 | Silent Mutation | CCC,CCG | P,P 118 | NP_002156.2 | |
NM_181353.2 | 459 | Silent Mutation | CCC,CCG | P,P 118 | NP_851998.1 |
MIR3193 - microRNA 3193 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |