Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607984 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRY4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPRY4 - sprouty RTK signaling antagonist 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127496.1 | 3513 | UTR 3 | NP_001120968.1 | |||
NM_001293289.1 | 3513 | UTR 3 | NP_001280218.1 | |||
NM_001293290.1 | 3513 | UTR 3 | NP_001280219.1 | |||
NM_030964.3 | 3513 | UTR 3 | NP_112226.2 | |||
XM_011537685.2 | 3513 | UTR 3 | XP_011535987.1 | |||
XM_017009910.1 | 3513 | UTR 3 | XP_016865399.1 |
SPRY4-IT1 - SPRY4 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |