Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCGGAGAGGGCACGTACGGAGTT[C/G]TGTACAAAGCCAGAAACAAGTTGAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 116953 MIM: 155550 MIM: 179514 | |||||||||||||||||||||||
Literature Links: |
CDK2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CDK2 - cyclin dependent kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290230.1 | 292 | Missense Mutation | CTG,GTG | L,V 18 | NP_001277159.1 | |
NM_001798.4 | 292 | Missense Mutation | CTG,GTG | L,V 18 | NP_001789.2 | |
NM_052827.3 | 292 | Missense Mutation | CTG,GTG | L,V 18 | NP_439892.2 | |
XM_011537732.1 | 292 | Missense Mutation | CTG,GTG | L,V 18 | XP_011536034.1 |
PMEL - premelanosome protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200053.1 | 292 | Intron | NP_001186982.1 | |||
NM_001200054.1 | 292 | Intron | NP_001186983.1 | |||
NM_001320121.1 | 292 | Intron | NP_001307050.1 | |||
NM_001320122.1 | 292 | Intron | NP_001307051.1 | |||
NM_006928.4 | 292 | Intron | NP_008859.1 | |||
XM_006719569.1 | 292 | Intron | XP_006719632.1 | |||
XM_011538685.1 | 292 | Intron | XP_011536987.1 |
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |