Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCCTATATTGTCTATGCCCAGA[C/T]GGTCTTGTAACAGAAAACAGGAATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616778 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP22 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DUSP22 - dual specificity phosphatase 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286555.1 | Intron | NP_001273484.1 | ||||
NM_020185.4 | Intron | NP_064570.1 | ||||
XM_011514757.2 | Intron | XP_011513059.1 | ||||
XM_017011061.1 | Intron | XP_016866550.1 | ||||
XM_017011062.1 | Intron | XP_016866551.1 |
Set Membership: |
HapMap |