Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACAGTCTTGGTTCCCCGGAGA[C/T]GTCCAGTCCGGCGGGCAGCAATGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604177 MIM: 194526 | ||||||||||||||||||||
Literature Links: |
MIR6850 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6850 - microRNA 6850 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL8 - ribosomal protein L8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000973.4 | 869 | Missense Mutation | CAT,CGT | H,R 245 | NP_000964.1 | |
NM_001317771.1 | 869 | Missense Mutation | CAT,CGT | H,R 245 | NP_001304700.1 | |
NM_001317782.1 | 869 | Missense Mutation | CAT,CGT | H,R 245 | NP_001304711.1 | |
NM_033301.2 | 869 | Missense Mutation | CAT,CGT | H,R 245 | NP_150644.1 | |
XM_017013723.1 | 869 | Intron | XP_016869212.1 |
ZNF34 - zinc finger protein 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF517 - zinc finger protein 517 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |