Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
74 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612317 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATAD3B - ATPase family, AAA domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317238.1 | 1698 | Silent Mutation | ATC,ATT | I,I 421 | NP_001304167.1 | |
NM_031921.5 | 1698 | Silent Mutation | ATC,ATT | I,I 467 | NP_114127.3 | |
XM_005244806.3 | 1698 | Silent Mutation | ATC,ATT | I,I 467 | XP_005244863.1 | |
XM_011542241.2 | 1698 | Intron | XP_011540543.1 | |||
XM_011542243.2 | 1698 | Silent Mutation | ATC,ATT | I,I 299 | XP_011540545.1 | |
XM_011542244.1 | 1698 | Silent Mutation | ATC,ATT | I,I 515 | XP_011540546.1 |