Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTTTTCCTTAGAGGGGCCACTT[A/C]GGCTTACACTTCCGGATCCTGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTR - 5-methyltetrahydrofolate-homocysteine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000254.2 | 780 | Intron | NP_000245.2 | |||
NM_001291939.1 | 780 | Intron | NP_001278868.1 | |||
NM_001291940.1 | 780 | Intron | NP_001278869.1 | |||
XM_005273141.4 | 780 | Intron | XP_005273198.1 | |||
XM_006711770.2 | 780 | Intron | XP_006711833.1 | |||
XM_011544194.2 | 780 | UTR 5 | XP_011542496.1 | |||
XM_017001329.1 | 780 | UTR 5 | XP_016856818.1 | |||
XM_017001330.1 | 780 | UTR 5 | XP_016856819.1 |