Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTCCATTGGTGGAAATATCAAA[C/T]TAACTCCTTGTGGTCCTGGGATAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613476 MIM: 616497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD6 - major facilitator superfamily domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017694.3 | 2568 | Intron | NP_060164.3 | |||
XM_005246654.2 | 2568 | Intron | XP_005246711.1 | |||
XM_005246656.3 | 2568 | Intron | XP_005246713.1 | |||
XM_011511368.1 | 2568 | Intron | XP_011509670.1 | |||
XM_017004392.1 | 2568 | Intron | XP_016859881.1 |
NEMP2 - nuclear envelope integral membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142645.1 | 2568 | Intron | NP_001136117.1 | |||
XM_005246212.3 | 2568 | Intron | XP_005246269.1 | |||
XM_005246213.3 | 2568 | Intron | XP_005246270.1 | |||
XM_011510458.2 | 2568 | UTR 3 | XP_011508760.1 | |||
XM_011510459.2 | 2568 | Intron | XP_011508761.1 | |||
XM_017003098.1 | 2568 | Intron | XP_016858587.1 | |||
XM_017003099.1 | 2568 | Intron | XP_016858588.1 | |||
XM_017003100.1 | 2568 | UTR 3 | XP_016858589.1 |