Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATCTGTACTTGGTAGAAACAAA[A/G]AAAGTCATGTAGAAAATACTAGTAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600355 | |||||||||||||||||||||||
Literature Links: |
NAIP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
NAIP - NLR family apoptosis inhibitory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004536.2 | Intron | NP_004527.2 | ||||
NM_022892.1 | Intron | NP_075043.1 | ||||
XM_011543413.2 | Intron | XP_011541715.1 | ||||
XM_017009490.1 | Intron | XP_016864979.1 |
Set Membership: |
HapMap |