Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCCCCCGGACAGTGGCTCTGA[C/T]GGCGTTACTGATGGTGCTGCTCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142880 MIM: 142858 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-DPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLA-DPA1 - major histocompatibility complex, class II, DP alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242524.1 | 163 | Intron | NP_001229453.1 | |||
NM_001242525.1 | 163 | Intron | NP_001229454.1 | |||
NM_033554.3 | 163 | Intron | NP_291032.2 |
HLA-DPB1 - major histocompatibility complex, class II, DP beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002121.5 | 163 | Missense Mutation | ACG,ATG | T,M 16 | NP_002112.3 |