Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGGGGAGTGGTAGGTCACTGT[A/C]AGGTGGAGAGGAGCAGGGAGGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRBA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRBA1 - KRAB-A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290187.1 | Intron | NP_001277116.1 | ||||
NM_032534.3 | Intron | NP_115923.2 | ||||
XM_011516636.2 | Intron | XP_011514938.1 | ||||
XM_011516637.2 | Intron | XP_011514939.1 | ||||
XM_011516638.2 | Intron | XP_011514940.1 | ||||
XM_011516639.2 | Intron | XP_011514941.1 | ||||
XM_011516640.2 | Intron | XP_011514942.1 | ||||
XM_011516641.2 | Intron | XP_011514943.1 | ||||
XM_011516642.2 | Intron | XP_011514944.1 | ||||
XM_011516643.2 | Intron | XP_011514945.1 |