Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCCTCTCTTCCCGAAGAACTG[G/T]ACGAAGACATCTCCACGACTAGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165196 | ||||||||||||||||||||
Literature Links: |
OPRK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OPRK1 - opioid receptor kappa 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000912.4 | 1464 | UTR 3 | NP_000903.2 | |||
NM_001282904.1 | 1464 | UTR 3 | NP_001269833.1 | |||
NM_001318497.1 | 1464 | Missense Mutation | TAC,TCC | Y,S 389 | NP_001305426.1 |