Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGGCACTCGTCATGAATTGAAG[A/G]ATGAAAAGCCATAGTCACAAGTCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125270 MIM: 607267 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALAD PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALAD - aminolevulinate dehydratase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C9orf43 - chromosome 9 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLE3 - DNA polymerase epsilon 3, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278255.1 | 1556 | UTR 3 | NP_001265184.1 | |||
NM_017443.4 | 1556 | UTR 3 | NP_059139.3 |
Set Membership: |
HapMap |