Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTCCCACAGTGCTCTGAGCGCC[A/G]TAGCCTCCCTCCTCCCCCCACAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125270 MIM: 607267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALAD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALAD - aminolevulinate dehydratase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C9orf43 - chromosome 9 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278629.1 | Intron | NP_001265558.1 | ||||
NM_001278630.1 | Intron | NP_001265559.1 | ||||
NM_152786.2 | Intron | NP_689999.1 | ||||
XM_005251885.3 | Intron | XP_005251942.1 | ||||
XM_011518477.1 | Intron | XP_011516779.1 | ||||
XM_011518478.1 | Intron | XP_011516780.1 | ||||
XM_011518479.2 | Intron | XP_011516781.1 |
POLE3 - DNA polymerase epsilon 3, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278255.1 | Intron | NP_001265184.1 | ||||
NM_017443.4 | Intron | NP_059139.3 |