Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGTCATCTTTAATGGCTGTATT[A/G]GGCTTGGCCTGAGACCAGTATTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138120 MIM: 605962 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPA5 - heat shock protein family A (Hsp70) member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005347.4 | 3789 | UTR 3 | NP_005338.1 |
RABEPK - Rab9 effector protein with kelch motifs | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174152.1 | 3789 | Intron | NP_001167623.1 | |||
NM_001174153.1 | 3789 | Intron | NP_001167624.1 | |||
NM_005833.3 | 3789 | Intron | NP_005824.2 | |||
XM_005251640.4 | 3789 | Intron | XP_005251697.1 | |||
XM_005251641.4 | 3789 | Intron | XP_005251698.1 | |||
XM_005251642.4 | 3789 | Intron | XP_005251699.1 | |||
XM_005251644.4 | 3789 | Intron | XP_005251701.1 | |||
XM_011518120.2 | 3789 | Intron | XP_011516422.1 | |||
XM_017014177.1 | 3789 | Intron | XP_016869666.1 | |||
XM_017014178.1 | 3789 | Intron | XP_016869667.1 | |||
XM_017014179.1 | 3789 | Intron | XP_016869668.1 | |||
XM_017014180.1 | 3789 | Intron | XP_016869669.1 | |||
XM_017014181.1 | 3789 | Intron | XP_016869670.1 | |||
XM_017014182.1 | 3789 | Intron | XP_016869671.1 | |||
XM_017014183.1 | 3789 | Intron | XP_016869672.1 |