Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCACTTTCTTCAATTGGGAGCA[A/G]CTGAAATAGTCAGAGATTTGAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RTKN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RTKN2 - rhotekin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282941.1 | Intron | NP_001269870.1 | ||||
NM_145307.3 | Intron | NP_660350.2 | ||||
XM_011539456.2 | Intron | XP_011537758.1 | ||||
XM_011539457.2 | Intron | XP_011537759.1 | ||||
XM_011539460.2 | Intron | XP_011537762.1 | ||||
XM_017015842.1 | Intron | XP_016871331.1 | ||||
XM_017015843.1 | Intron | XP_016871332.1 | ||||
XM_017015844.1 | Intron | XP_016871333.1 |