Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGTCTTACCTGGAAGAGGAG[C/T]GGAGTTTGCAGCATCAAGTCCTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALCA - calcitonin related polypeptide alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033952.2 | 1074 | Intron | NP_001029124.1 | |||
NM_001033953.2 | 1074 | Intron | NP_001029125.1 | |||
NM_001741.2 | 1074 | Intron | NP_001732.1 | |||
XM_017018283.1 | 1074 | UTR 3 | XP_016873772.1 | |||
XM_017018284.1 | 1074 | UTR 3 | XP_016873773.1 |