Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACACGCGCGGGGGCCGGAACTGC[C/G]GTCGCCGGCGCGGTCGTTGTCGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615910 | ||||||||||||||||||||
Literature Links: |
NAXD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAXD - NAD(P)HX dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242881.1 | 121 | Silent Mutation | GCC,GCG | A,A 10 | NP_001229810.1 | |
NM_001242882.1 | 121 | Intron | NP_001229811.1 | |||
NM_001242883.1 | 121 | Intron | NP_001229812.1 | |||
NM_018210.3 | 121 | Silent Mutation | GCC,GCG | A,A 10 | NP_060680.2 |