Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGTTGCTGCTGTTGTTGCTG[C/T]TGTTGTTGGTGCTGTTGCTGCTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609280 MIM: 600708 | ||||||||||||||||||||
Literature Links: |
EIF2AK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF2AK4 - eukaryotic translation initiation factor 2 alpha kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013703.3 | 465 | Intron | NP_001013725.2 | |||
XM_005254392.2 | 465 | Intron | XP_005254449.1 | |||
XM_011521599.2 | 465 | Intron | XP_011519901.1 | |||
XM_011521600.2 | 465 | Intron | XP_011519902.1 | |||
XM_017022219.1 | 465 | Intron | XP_016877708.1 |
SRP14 - signal recognition particle 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001309434.1 | 465 | Missense Mutation | ACA,GCA | T,A 79 | NP_001296363.1 | |
NM_003134.5 | 465 | Missense Mutation | ACA,GCA | T,A 127 | NP_003125.3 |
SRP14-AS1 - SRP14 antisense RNA1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |