Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCCTCTTAGTGATCAATCATCT[C/G]TCACGCAAAAATTCACTCTCTTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600178 MIM: 605230 | ||||||||||||||||||||
Literature Links: |
MAP1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP1A - microtubule associated protein 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002373.5 | Intron | NP_002364.5 | ||||
XM_017022189.1 | Intron | XP_016877678.1 |
TP53BP1 - tumor protein p53 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |