Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCCAGAATGAGCTCAGCTGAC[A/G]TCTCCTTGGAAGAGAGGGAGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603687 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH1A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH1A2 - aldehyde dehydrogenase 1 family member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206897.1 | 2260 | UTR 3 | NP_001193826.1 | |||
NM_003888.3 | 2260 | UTR 3 | NP_003879.2 | |||
NM_170696.2 | 2260 | UTR 3 | NP_733797.1 | |||
NM_170697.2 | 2260 | UTR 3 | NP_733798.1 |