Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGCAGCTTCCTCCAAACAAGTA[C/T]AGGTAGGGGTTGAGACCCAGGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124095 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSK - c-src tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127190.1 | Intron | NP_001120662.1 | ||||
NM_004383.2 | Intron | NP_004374.1 | ||||
XM_005254165.4 | Intron | XP_005254222.1 | ||||
XM_017021925.1 | Intron | XP_016877414.1 |
MIR4513 - microRNA 4513 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |