Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTGCCAGGGGACTGGGGAAGTG[A/G]GGCTCCTGACCACTCATCTCACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARRB2 - arrestin beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257328.1 | Intron | NP_001244257.1 | ||||
NM_001257329.1 | Intron | NP_001244258.1 | ||||
NM_001257330.1 | Intron | NP_001244259.1 | ||||
NM_001257331.1 | Intron | NP_001244260.1 | ||||
NM_004313.3 | Intron | NP_004304.1 | ||||
NM_199004.1 | Intron | NP_945355.1 | ||||
XM_006721520.1 | Intron | XP_006721583.1 | ||||
XM_011523858.2 | Intron | XP_011522160.1 | ||||
XM_017024645.1 | Intron | XP_016880134.1 |