Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCAAATTAAATTTAGCAGAGTT[A/T]ATTTGAGCAAAGAAGTGACTCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2B6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP2B6 - cytochrome P450 family 2 subfamily B member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000767.4 | Intron | NP_000758.1 | ||||
XM_005258569.4 | Intron | XP_005258626.1 | ||||
XM_006723050.3 | Intron | XP_006723113.1 | ||||
XM_011526546.2 | Intron | XP_011524848.1 | ||||
XM_011526547.2 | Intron | XP_011524849.1 | ||||
XM_011526548.2 | Intron | XP_011524850.1 | ||||
XM_011526549.2 | Intron | XP_011524851.1 | ||||
XM_011526550.2 | Intron | XP_011524852.1 |