Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAATCCTCTGCCCCGCGGCGCAG[C/T]CGAGCCCCGAGCCAGGAGTCGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD15 - potassium channel tetramerization domain containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129994.1 | Intron | NP_001123466.1 | ||||
NM_001129995.1 | Intron | NP_001123467.1 | ||||
NM_024076.2 | Intron | NP_076981.2 | ||||
XM_011527296.1 | Intron | XP_011525598.1 | ||||
XM_011527297.2 | Intron | XP_011525599.1 | ||||
XM_011527298.2 | Intron | XP_011525600.1 | ||||
XM_017027283.1 | Intron | XP_016882772.1 | ||||
XM_017027284.1 | Intron | XP_016882773.1 |