Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTAGGCAGCCCACAAGGCCCTT[G/T]GTGATGTTATTTCTGCCTTCTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615321 | ||||||||||||||||||||
Literature Links: |
CLIC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLIC6 - chloride intracellular channel 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317009.1 | Intron | NP_001303938.1 | ||||
NM_053277.2 | Intron | NP_444507.1 | ||||
XM_017028406.1 | Intron | XP_016883895.1 |