Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTGCTGATGGAGTTGGGGAAA[C/T]TGGCTGAAGAAAAAGTGAGAAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC14L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC14L2 - SEC14 like lipid binding 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204204.2 | Intron | NP_001191133.1 | ||||
NM_001291932.1 | Intron | NP_001278861.1 | ||||
NM_012429.4 | Intron | NP_036561.1 | ||||
NM_033382.2 | Intron | NP_203740.1 |